Learn More
Invitrogen™ NCK2 Polyclonal Antibody

Description
The antibody detects endogenous level of total NCK2 protein.
Specifications
Specifications
| Antigen | NCK2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | NCK2 |
| Gene Accession No. | O43639, O55033 |
| Gene Alias | 4833426I10Rik; cytoplasmic protein NCK2; Grb4; growth factor receptor-bound protein 4; NCK adaptor protein 2; NCK2; Nck-2; NCKbeta; non-catalytic region of tyrosine kinase adaptor protein 2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta |
| Gene Symbols | NCK2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.