Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCKAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100161
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100161 Supplier Invitrogen™ Supplier No. PA5100161
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NCKAP1.

This gene interacts with Nck and may be involved in the downstream signaling of Nck and shares similarity with the mouse Dhm1 and the yeast dhp1 gene. The yeast gene is involved in homologous recombination and RNA metabolism, such as RNA synthesis and RNA trafficking. Complementation studies show that Dhm1 has a similar function in mouse as dhp1. The function of the human gene has not yet been determined. Transcript variants encoding different isoforms have been noted for this gene; however, their full-length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCKAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NCKAP1
Gene Accession No. P28660, P55161, Q9Y2A7
Gene Alias Brain protein H19; C79304; H19; HEM2; Hem-2; KIAA0587; Membrane-associated protein HEM-2; MH19; mKIAA0587; NAP 1; Nap1; NAP125; NCK associated protein 1; NCKAP1; NCK-associated protein 1; p125Nap1
Gene Symbols NCKAP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NCKAP1(Accession Q9Y2A7), corresponding to amino acid residues V863-I913.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10787, 50884, 58823
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.