Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCKAP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548958
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548958 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548958 Supplier Invitrogen™ Supplier No. PA548958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCKAP1L Polyclonal Antibody for Western Blot

This gene encodes a member of the HEM family of tissue-specific transmembrane proteins which are highly conserved from invertebrates through mammals. This gene is only expressed in hematopoietic cells. The encoded protein is a part of the Scar/WAVE complex which plays an important role in regulating cell shape in both metazoans and plants. Alternatively spliced transcript variants encoding different isoforms have been found.[provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCKAP1L
Applications Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NCKAP1L
Gene Accession No. P55160, Q8K1X4
Gene Alias 4930568P13Rik; AI463083; HEM1; Hematopoietic protein 1; Hemp1; membrane-associated protein hem-1; NCK associated protein 1 like; NCKAP1L; nck-associated protein 1-like
Gene Symbols NCKAP1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 641-669 amino acids from the Central region of human NCKPL.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105855, 3071
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.