Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCKAP5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983084
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3084 Supplier Bioss Supplier No. BS13693R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCKAP5 (NCK Associated Protein 5) is a Protein Coding gene. An important paralog of this gene isNCKAP5L.

Specifications

Antigen NCKAP5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NCKAP5
Gene Accession No. O14513
Gene Alias 8430408F21; D130011D22Rik; E030049G20Rik; ERIH; ERIH1; Erih2; Gm1548; NAP5; NAP-5; NCK associated protein 5; NCKAP5; Nck-associated protein 5; Peripheral clock protein; peripheral clock protein 2; RGD1564507
Gene Symbols NCKAP5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NCKAP5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344148
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.