Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCKAP5L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559404
Description
Immunogen sequence: EEKVMKGIEE NVLRLQGQER APGAEVKHRN TSSIASWFGL KKSKLPALNR RTEATKNKEG AGGGSPLRRE VKMEARKL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
KIAA1602, also named as NCKAP5L, has 4 isoforms. The target band is about 139-145 kDa for phosphorylation.
Specifications
NCKAP5L | |
Polyclonal | |
Unconjugated | |
NCKAP5L | |
C230021P08Rik; Centrosomal protein of 169 kDa; Cep169; FP1193; KIAA1602; NCK associated protein 5 like; NCKAP5L; NCKAP5-like; NCK-associated protein 5-like; RGD1561571 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57701 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCH0 | |
NCKAP5L | |
Recombinant protein corresponding to Human NCKAP5L. Recombinant protein control fragment (Product #RP-97809). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction