Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCKIPSD Monoclonal Antibody (2E4)
GREENER_CHOICE

Catalog No. PIMA521948
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521948 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521948 Supplier Thermo Scientific Supplier No. MA521948
Add to Cart
Add to Cart

NCKIPSD Monoclonal Antibody (2E4)

NCKIPSD Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen NCKIPSD
Applications ELISA
Classification Monoclonal
Clone 2E4
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene NCKIPSD
Gene Accession No. Q9NZQ3
Gene Alias 54 kDa VacA-interacting protein; 54 kDa vimentin-interacting protein; 90 kDa N-WASP-interacting protein; 90 kDa SH3 protein interacting with Nck; AF3P21; dia interacting protein; dia-interacting protein 1; diaphanous protein interacting protein; Diaphanous protein-interacting protein; DIP; DIP1; DIP-1; mDia interacting protein-1; MGC23891; NCK interacting protein with SH3 domain; NCK-interacting protein with SH3 domain; Nckipsd; N-WASP-binding protein; ORF1; SH3 adapter protein SPIN90; SH3 protein interacting with Nck, 90 kDa; Spin90; VIP54; Wasbp; WASLBP; WASP-interacting SH3-domain protein; WISH; Wiskott-Aldrich syndrome homolog binding protein; Wiskott-Aldrich syndrome protein-binding protein; wiskott-Aldrich syndrome protein-binding protein WISH; wiskott-Aldrich syndrome protein-interacting protein
Gene Symbols NCKIPSD
Host Species Mouse
Immunogen NCKIPSD (NP_909119.1, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51517
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.