Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCKIPSD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530625 Supplier Invitrogen™ Supplier No. PA530625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116, mouse brain. Predicted reactivity: Mouse (91%), Rhesus Monkey (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is localized exclusively in the cell nucleus. It plays a role in signal transduction, and may function in the maintenance of sarcomeres and in the assembly of myofibrils into sarcomeres. It also plays an important role in stress fiber formation. The gene is involved in therapy-related leukemia by a chromosomal translocation t(3;11)(p21;q23) that involves this gene and the myeloid/lymphoid leukemia gene. Alternative splicing occurs in this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCKIPSD
Applications Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene NCKIPSD
Gene Accession No. Q9ESJ4, Q9NZQ3
Gene Alias 54 kDa VacA-interacting protein; 54 kDa vimentin-interacting protein; 90 kDa N-WASP-interacting protein; 90 kDa SH3 protein interacting with Nck; AF3P21; dia interacting protein; dia-interacting protein 1; diaphanous protein interacting protein; Diaphanous protein-interacting protein; DIP; DIP1; DIP-1; mDia interacting protein-1; MGC23891; NCK interacting protein with SH3 domain; NCK-interacting protein with SH3 domain; Nckipsd; N-WASP-binding protein; ORF1; SH3 adapter protein SPIN90; SH3 protein interacting with Nck, 90 kDa; Spin90; VIP54; Wasbp; WASLBP; WASP-interacting SH3-domain protein; WISH; Wiskott-Aldrich syndrome homolog binding protein; Wiskott-Aldrich syndrome protein-binding protein; wiskott-Aldrich syndrome protein-binding protein WISH; wiskott-Aldrich syndrome protein-interacting protein
Gene Symbols NCKIPSD
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human NCKIPSD. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51517, 80987
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.