Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
NCL 00017509 is a potent and reversible NIMA related kinase 2 (Nek2) inhibitor (IC50 = 56 nM). Induces increased mitotic abnormalities and mitotic delay. Cell permeable and active in vivo. In mice bearing pancreatic tumors, NCL 00017509 decreases PD-L1 expression and induces an anticancer immune response.
Specifications
Specifications
| Quantity | 1 mg |
| Solubility | Soluble to 100mM in DMSO |
| Formula Weight | 292.3 |
| Physical Form | Solid |
| Purity | >97% |
| Molecular Weight (g/mol) | 292.302 |
| Chemical Name or Material | 3-[(6-Ethynyl-9H-purin-2-yl)amino]benzeneacetamide |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

