Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCLN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104276
Description
Antibody detects endogenous levels of total Nicalin.
May antagonize Nodal signaling and subsequent organization of axial structures during mesodermal patterning. [UniProt].
Specifications
NCLN | |
Polyclonal | |
Unconjugated | |
Ncln | |
3100002P13Rik; AW120537; NCLN; NET59; Nicalin; nicalin homolog; nicalin homolog (zebrafish); nicastrin-like protein; RGD1309355 | |
Rabbit | |
Affinity chromatography | |
RUO | |
103425, 314648, 56926 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5XIA1, Q8VCM8, Q969V3 | |
Ncln | |
A synthesized peptide derived from human NCLN(Accession Q969V3), corresponding to amino acid residues A34-C84. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction