Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCLN Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570891
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-157-0891 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0891 Supplier Bethyl Laboratories, Inc Supplier No. A305623AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Nicalin, along with its binding partner Nomo, is a novel signaling antagonist to Nodals, signaling factors of the TGF-Beta superfamily that play a key role in vertebrate development. Nicalin is distantly related to Nicastrin, a component of the Alzheimer′s disease-associated gamma-secretase, and is thought to have a similar function. Another protein, TMEM147, has also been recently identified as associating with Nicalin and NOMO as part of the gamma-secretase-like complex.

Specifications

Antigen NCLN
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene NCLN
Gene Accession No. Q8VCM8, Q969V3
Gene Alias NET59, nicalin homolog (zebrefish), nicastrin-like protein
Gene Symbols Ncln
Host Species Rabbit
Immunogen Between 375 and 425
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103425, 56926
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.