Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NCoA-2, Mouse, Unlabeled, Clone: 1C6.3,

Catalog No. MABS1688MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1688MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1688MI Supplier MilliporeSigma™ Supplier No. MABS1688
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nuclear receptor coactivator 2 (UniProt Q15596; also known as bHLHe75, Class E basic helix-loop-helix protein 75, hTIF2, NCoA-2, Transcriptional intermediary factor 2) is encoded by the NCOA2 (also known as BHLHE75, SRC2, TIF2) gene (Gene ID 10499) in human. Nuclear receptor coactivator 2 (NCoA-2) is a 160kDa transcriptional coactivator for steroid receptors and nuclear receptors that acts as a coactivator of the steroid binding domain (AF-2), but not the modulating N-terminal domain (AF-1). It possesses a single NR interaction domain (NID) and two autonomous activation domains, AD1 and AD2. NCoA-2 assists nuclear receptors in the upregulation of DNA expression. In cells ligand activated nuclear receptors recruit NCoA-2 to DNA where it acylates histones to make targeted regions accessible to transcriptional machinery. Along with NCOA1 it controls energy balance between white and brown adipose tissues and also acts as a critical regulator of glucose metabolism. It is also involved in the positive regulation of the transcriptional activity of the glucocorticoid receptor NR3C1 by sumoylation enhancer RWDD3. NCoA-2 is reported to positively regulate the circadian clock by acting as a transcriptional coactivator for the CLOCK-ARNTL/BMAL1 heterodimer. Chromosomal aberrations involving NCOA2 may be a cause of acute myeloid leukemias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCoA-2
Applications Western Blot
Classification Monoclonal
Clone 1C6.3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2a in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene NCOA2, BHLHE75, SRC2, TIF2
Gene Accession No. Q15596
Host Species Mouse
Immunogen GST-tagged recombinant human NCoA-2 C-terminal region fragment..
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_059111
Test Specificity Clone 1C6.3 targeted epitope lies within the C-terminal one fourth region of the full-length human NCoA-2.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.