Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOA2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100388JF646
Description
NCOA2 Polyclonal antibody specifically detects NCOA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
NCOA2 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
BHLHE75, bHLHe75MGC138808, Class E basic helix-loop-helix protein 75, EC 2.3.1.48, GRIP1glucocorticoid receptor-interacting protein-1, KAT13C, NCoA-2SRC2, nuclear receptor coactivator 2, TIF2hTIF2, Transcriptional intermediary factor 2 | |
The immunogen recognized by this antibody maps to a region between residues 700 and 750 of human Nuclear Receptor Coactivator 2 using the numbering given in entry NP_006531.1 (GeneID 10499). | |
0.1 mL | |
Cancer, DNA Repair, Transcription Factors and Regulators | |
10499 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction