Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NCOA2 Split FISH Probe

Catalog No. p-7190149 Shop All Abnova Corporation Products
Click to view available options
Quantity:
200 μL

Includes

DAPI Counterstain (1500ng/mL) 250μL

Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.

FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.

Advantages

  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity

Specifications

Content And Storage Store at 4°C in the dark.
Label or Dye FITC, Texas Red
For Use With (Application) Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
Includes DAPI Counterstain (1500 ng/mL ) 250 uL
Location 8q13.3, 8q13.3
Probe NCOA2, NCOA2
Product Type NCOA2 Split FISH Probe
Quantity 200 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.