Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOA3/AIB1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100314B
Description
NCOA3/AIB1 Polyclonal antibody specifically detects NCOA3/AIB1 in Human samples. It is validated for Western BlotSpecifications
NCOA3/AIB1 | |
Polyclonal | |
Western Blot | |
ACTRCBP-interacting protein, AIB1AIB-1, Amplified in breast cancer 1 protein, BHLHE42, bHLHe42p/CIP, CAGH16, Class E basic helix-loop-helix protein 42, CTG26, KAT13B, MGC141848, NCoA-3, nuclear receptor coactivator 3, pCIP, RAC-3, RAC3pCIP, Receptor-associated coactivator 3, SRC3, SRC-3EC 2.3.1.48, Steroid receptor coactivator protein 3, Thyroid hormone receptor activator molecule 1, TNRC16, TRAM1, TRAM-1TNRC14 | |
A synthetic peptide, which represented a portion of human nuclear receptor coactivator 3 encoded within exon 13 (LocusLink ID 8202). | |
0.1 mL | |
Breast Cancer, Cancer, Chromatin Research, Transcription Factors and Regulators | |
8202 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction