Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOA4 Antibody (OTI2G4), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272895AF647
Description
NCOA4 Monoclonal specifically detects NCOA4 in Human samples. It is validated for Western Blot.Specifications
NCOA4 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NCOA4 | |
Recombinant protein fragment corresponding to amino acids 280-553 of human NCOA4 (NP_001138732) produced in E.coli. | |
0.1 mL | |
8031.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2G4 | |
Western Blot | |
Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction