Learn More
NCOA4 Mouse anti-Human, Clone: OTI2G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806402

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Androgen receptor (AR) coactivator ARA70, also designated NCOA4, RFG and ELE1, is a putative co-activator that specifically enhances the activity of the androgen receptor. In human thyroid carcinomas, the Ret proto-oncogene fuses to ARA70 to form Ret/PTC3 by an intrachromosomal inversion of chromosome in vivo. ARA70 is expressed as two isoforms, ARA70a and ARA70b. The shorter variant, ARA70b, results from an internal 985-bp deletion. ARA70a is widely expressed, and its expression is highest in testis and adipose tissues, whereas ARA70b is solely expressed in the testis. ARA70a can function as a ligand-enhanced co-activator of PPARg in adipocytes. ARA70 may be involved in prostate carcinogenesis and ovarian cancer and may serve as a key mediator of estrogen-androgen synergism.Specifications
NCOA4 | |
Monoclonal | |
Unconjugated | |
NCOA4 | |
ARA70, ELE1, PTC3, RFG | |
Mouse | |
Affinity Chromatography | |
RUO | |
8031 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2G4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13772 | |
NCOA4 | |
Human recombit protein fragment corresponding to amino acids 280-553 of human NCOA4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.