Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOA4 Polyclonal Antibody

NCOA4 Polyclonal Antibody
Supplier: Thermo Scientific PA5115626
Description
NCOA4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

Specifications
NCOA4 | |
Polyclonal | |
Unconjugated | |
NCOA4 | |
70 kDa androgen receptor coactivator; 70 kDa AR-activator; Androgen receptor coactivator 70 kDa protein; Androgen receptor-associated protein of 70 kDa; ARA70; ELE1; NCOA4; NCoA-4; Nuclear receptor coactivator 4; PTC3; ret fused; RET-activating gene ELE1; Ret-activating protein ELE1; RFG | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27057, 619385, 8031 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q13772 | |
NCOA4 | |
A synthesized peptide derived from human NCOA4(Accession Q13772), corresponding to amino acid residues I499-L549. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction