Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCOA5 Monoclonal Antibody (1E9)
GREENER_CHOICE

Catalog No. PIMA522408
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522408 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522408 Supplier Invitrogen™ Supplier No. MA522408
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAGMPGTAET FETPETCGTT DMRDSRDPMY RREGSYDRYL RMDDYCRRKD DSYFDRYRDS FDGRGPPGPE SQSRAKERLK REERRREELY RQYFEEIQRR FDAERPVDCS VIVVNKQTKD YAESVGRKVR DLGMVVDLIF LNTEVSLSQA LEDVSRGGSP FAIVITQQHQ IHRSCTVNIM FGTPQEHRNM PQADAMVLVA RNYERYKNEC REKEREEIAR QAAKMADEAI LQERERGGPE EGVRGGHPPA IQSLINLLAD NRYLTAEETD KIINYLRERK ERLMRSSTDS LPGELRGRAE ARFPANHSGR PRVPR.

This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Several alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCOA5
Applications ELISA
Classification Monoclonal
Clone 1E9
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene NCOA5
Gene Accession No. Q9HCD5
Gene Alias bA465L10.6; CIA; coactivator independent of AF-2; KIAA1637; NCOA5; NCoA-5; Nuclear receptor coactivator 5
Gene Symbols NCOA5
Host Species Mouse
Immunogen NCOA5 (AAH56872, 1 a.a. ~ 315 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57727
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.