Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCOA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562156 Supplier Invitrogen™ Supplier No. PA562156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NECREKEREE IARQAAKMAD EAILQERERG GPEEGVRGGH PPAIQSLINL LADNRYLTAE ETDKIINYLR ERKERLMRSS TDSLPGPISR QPL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Several alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCOA5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCOA5
Gene Accession No. Q9HCD5
Gene Alias bA465L10.6; CIA; coactivator independent of AF-2; KIAA1637; NCOA5; NCoA-5; Nuclear receptor coactivator 5
Gene Symbols NCOA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NCOA5. Recombinant protein control fragment (Product #RP-99337).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57727
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.