Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA6 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB256062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB256062 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB256062 Supplier Novus Biologicals Supplier No. NB200336V
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCOA6 Polyclonal antibody specifically detects NCOA6 in Human samples. It is validated for Western Blot

Specifications

Antigen NCOA6
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Activating signal cointegrator 2, AIB3NRC RAP250, Amplified in breast cancer protein 3, ASC-2, Cancer-amplified transcriptional coactivator ASC-2, KIAA0181PPAR-interacting protein, NRCamplified in breast cancer-3 protein, nuclear receptor coactivator 6, Nuclear receptor coactivator RAP250, Nuclear receptor-activating protein, 250 kDa, peroxisome proliferator-activated receptor interacting protein, PRIPPeroxisome proliferator-activated receptor-interacting protein, RAP250Thyroid hormone receptor-binding protein, thyroid hormone receptor binding protein, TRBPASC2activating signal cointegrator-2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2000 and the C-terminus (residue 2063) of human Activating Signal Cointegrator-2 using the numbering given in Swiss-Prot entry Q14686 (GeneID 23054).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 23054
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.