Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOA6 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200336H
Description
NCOA6 Polyclonal antibody specifically detects NCOA6 in Human samples. It is validated for Western BlotSpecifications
NCOA6 | |
Polyclonal | |
Western Blot | |
Activating signal cointegrator 2, AIB3NRC RAP250, Amplified in breast cancer protein 3, ASC-2, Cancer-amplified transcriptional coactivator ASC-2, KIAA0181PPAR-interacting protein, NRCamplified in breast cancer-3 protein, nuclear receptor coactivator 6, Nuclear receptor coactivator RAP250, Nuclear receptor-activating protein, 250 kDa, peroxisome proliferator-activated receptor interacting protein, PRIPPeroxisome proliferator-activated receptor-interacting protein, RAP250Thyroid hormone receptor-binding protein, thyroid hormone receptor binding protein, TRBPASC2activating signal cointegrator-2 | |
The immunogen recognized by this antibody maps to a region between residue 2000 and the C-terminus (residue 2063) of human Activating Signal Cointegrator-2 using the numbering given in Swiss-Prot entry Q14686 (GeneID 23054). | |
0.1 mL | |
Apoptosis, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
23054 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction