Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCOA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556430 Supplier Invitrogen™ Supplier No. PA556430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HGSPTVTKLS KEPSDTSAAF ESTAKENFLG EDDDFVDLEE LSSQTGGGMH KKDTLKECLS LDPEERKKAE SQINNSAVEM QVQS Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 55%.

NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCOA7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCOA7
Gene Accession No. Q8NI08
Gene Alias 140 kDa estrogen receptor-associated protein; 9030406N13Rik; dJ187J11.3; ERAP140; ESNA1; Estrogen nuclear receptor coactivator 1; estrogen receptor associated protein 140 kDa; Nbla00052; Nbla10993; Ncoa7; NCOA7-AS; Nuclear receptor coactivator 7; putative protein product of Nbla00052; putative protein product of Nbla10993; RGD1566426; TBC/LysM-associated domain containing 4; TLDC4
Gene Symbols NCOA7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NCOA7. Recombinant protein control fragment (Product #RP-94661).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.