Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOR1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297461PCP
Description
NCOR1 Polyclonal antibody specifically detects NCOR1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
NCOR1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
hN-CoR, KIAA1047N-CoR1, MGC104216, N-CoRhCIT529I10, nuclear receptor corepressor 1, nuclear receptor co-repressor 1, thyroid hormone- and retinoic acid receptor-associated corepressor 1, TRAC1 | |
Produced in rabbits immunized with E. coli-derived Human NCOR1 fragment. | |
0.1 mL | |
Transcription Factors and Regulators | |
9611 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction