Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOR1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10058824 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10058824 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10058824 Supplier Novus Biologicals Supplier No. NB10058824
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

NCOR1 Polyclonal specifically detects NCOR1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation, Chromatin Immunoprecipitation Sequencing.

Specifications

Antigen NCOR1
Applications Western Blot, Immunoprecipitation, ChIP Assay
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate, Chromatin Immunoprecipitation (ChIP) 10 ug, Chromatin Immunoprecipitation Sequencing 2 ug
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O75376
Gene Alias hN-CoR, KIAA1047N-CoR1, MGC104216, N-CoRhCIT529I10, nuclear receptor corepressor 1, nuclear receptor co-repressor 1, thyroid hormone- and retinoic acid receptor-associated corepressor 1, TRAC1
Gene Symbols NCOR1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1525 and 1575 of human nuclear receptor co-repressor 1 using the numbering given in entry NP_006302.2 (GeneID 9611).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9611
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Duckbill platypus, White-tuftedear marmoset, Crab-eating macaque, European domestic ferret and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.