Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCoR1 Monoclonal Antibody (A6H12)
GREENER_CHOICE

Catalog No. PIMA544982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544982 Supplier Invitrogen™ Supplier No. MA544982
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NCoR1 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a protein that mediates ligand-independent transcription repression of thyroid-hormone and retinoic-acid receptors by promoting chromatin condensation and preventing access of the transcription machinery. It is part of a complex which also includes histone deacetylases and transcriptional regulators similar to the yeast protein Sin3p. This gene is located between the Charcot-Marie-Tooth and Smith-Magenis syndrome critical regions on chromosome 17. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 17 and 20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCoR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A6H12
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Ncor1
Gene Accession No. O75376, Q60974, Q9WUB5
Gene Alias 5730405M06Rik; A230020K14Rik; h KIAA1047; hCIT529I10; hN-CoR; KIAA1047; MGC104216; mKIAA1047; NCoR; N-CoR; N-Cor/SMRT corepressor Rip13; N-Cor/SMRT corepressor, Rip13; NCOR1; N-CoR1; nuclear receptor corepressor; nuclear receptor corepressor 1; nuclear receptor co-repressor 1; PPP1R109; protein phosphatase 1, regulatory subunit 109; retinoid X receptor interacting protein 13; retinoid X receptor-interacting protein 13; RIP13; Rxrip13; thyroid hormone- and retinoic acid receptor-associated corepressor 1; TRAC1
Gene Symbols Ncor1
Host Species Mouse
Immunogen Recombinant protein within human Nuclear Receptor Corepressor aa 251-450.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20185, 54299, 9611
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.