Learn More
Invitrogen™ NCoR1 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Specifications
Specifications
| Antigen | NCoR1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Ncor1 |
| Gene Accession No. | O75376 |
| Gene Alias | 5730405M06Rik; A230020K14Rik; h KIAA1047; hCIT529I10; hN-CoR; KIAA1047; MGC104216; mKIAA1047; NCoR; N-CoR; N-Cor/SMRT corepressor Rip13; N-Cor/SMRT corepressor, Rip13; NCOR1; N-CoR1; nuclear receptor corepressor; nuclear receptor corepressor 1; nuclear receptor co-repressor 1; PPP1R109; protein phosphatase 1, regulatory subunit 109; retinoid X receptor interacting protein 13; retinoid X receptor-interacting protein 13; RIP13; Rxrip13; thyroid hormone- and retinoic acid receptor-associated corepressor 1; TRAC1 |
| Gene Symbols | Ncor1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.