Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOR2 Antibody (1542), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202781
Description
NCOR2 Monoclonal specifically detects NCOR2 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
NCOR2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 4 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
NCOR2 | |
Synthetic peptide corresponding to residues C G G A(2016) S A S D P H R E K T Q(2027) of human NCOR2. | |
100 μg | |
Cancer, Chromatin Research, Transcription Factors and Regulators | |
9612 | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen), ChIP Assay | |
1542 | |
Unconjugated | |
CTG repeat protein 26, CTG26nuclear receptor co-repressor 2, FLJ25011, N-CoR2, nuclear receptor corepressor 2, retinoic-acid-receptor-associated corepressor, silencing mediator for retinoid and thyroid hormone receptors, SMAP270, SMRTE, SMRTE-tau, SMRTT3 receptor-associating factor, TNRC14, TRAC, TRAC1 | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
NCoR2 - CHIP Grade (1542) | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction