Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOR2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298866AF488
Description
NCOR2 Polyclonal specifically detects NCOR2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NCOR2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
CTG repeat protein 26, CTG26nuclear receptor co-repressor 2, FLJ25011, N-CoR2, nuclear receptor corepressor 2, retinoic-acid-receptor-associated corepressor, silencing mediator for retinoid and thyroid hormone receptors, SMAP270, SMRTE, SMRTE-tau, SMRTT3 receptor-associating factor, TNRC14, TRAC, TRAC1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human NCOR2. | |
0.1 mL | |
Cancer, Chromatin Research, Transcription Factors and Regulators | |
9612 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction