Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110855 Supplier Invitrogen™ Supplier No. PA5110855
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YITRNEMLDI VDAIYQMVGN TVELPEEENT PEKRVDRIFA MMDKNADGKL TLQEFQEGSK ADPSIVQALS LYDGLV Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ncs1
Gene Accession No. P62166, Q8BNY6
Gene Alias 9430075O15Rik; A730032G13Rik; AI836659; FLUP; Freq; frequenin; Frequenin homolog; frequenin-like protein; Frequenin-like ubiquitous protein; Mfreq; Ncs1; NCS-1; Neuronal calcium sensor 1; neuronal calcium sensor-1
Gene Symbols Ncs1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human NCS1. Recombinant protein control fragment (Product #RP-92904).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14299, 23413
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.