Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 105062AP150UL

Description
This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predomitly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
NCS1 | |
Polyclonal | |
Unconjugated | |
NCS1 | |
FLUP, FREQ, Frequenin homolog, frequenin homolog (Drosophila), Frequenin like protein, NCS 1, NCS1, Neuronal calcium sensor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14299, 23413, 65153 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62166, P62168, Q8BNY6 | |
Ncs1 | |
NCS1 Fusion Protein Ag0784 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction