Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NCS1 Recombinant Rabbit Monoclonal Antibody (JE56-39)
GREENER_CHOICE

Catalog No. PIMA544903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544903 Supplier Invitrogen™ Supplier No. MA544903
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NCS1 Recombinant Monoclonal Antibody for Western Blot

This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCS1
Applications Western Blot
Classification Recombinant Monoclonal
Clone JE56-39
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Ncs1
Gene Accession No. P62166, P62168, Q8BNY6
Gene Alias 9430075O15Rik; A730032G13Rik; AI836659; FLUP; Freq; frequenin; Frequenin homolog; frequenin-like protein; Frequenin-like ubiquitous protein; Mfreq; Ncs1; NCS-1; Neuronal calcium sensor 1; neuronal calcium sensor-1
Gene Symbols Ncs1
Host Species Rabbit
Immunogen Recombinant protein within human NCS1 aa 90-190.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14299, 23413, 65153
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.