Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NCT 501 hydrochloride
GREENER_CHOICE

Catalog No. 593410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-341-0 10 mg
59-345-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-341-0 Supplier Tocris Bioscience™ Supplier No. 5934/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ALDH1A1 inhibitor

NCT 501 hydrochloride is a potent and selective ALDH1A1 inhibitor (IC50 = 40 nM). Exhibits >1000-fold selectivity for ALDH1A1 over ALDH3A1, ALDH1B1, ALDH2, HPGD and HSD17β4. Also selective over a panel of 168 GPCRs. Blood-brain barrier permeable.

Specifications

Chemical Name or Material 8-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]methyl]-3,7-dihydro-1,3-dimethyl-7-(3-methylbutyl)-1H-purine-2,6-dione hydrochloride
CAS 1307896-46-3
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Molecular Formula C21H32N6O3.HCl
Formula Weight 452.98
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.