Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCU-G1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP275094 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275094 Supplier Novus Biologicals Supplier No. NBP275094
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

NCU-G1 Polyclonal specifically detects NCU-G1 in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen NCU-G1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Peptide ELISA
Formulation TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide
Gene Alias C1orf85, chromosome 1 open reading frame 85, lysosomal protein NCU-G1, MGC31963, NCU-G1, PSEC0030, UNQ2553/PRO6182
Gene Symbols C1orf85
Host Species Goat
Immunogen Peptide with sequence PDCPSVNERNSIDD, from the internal region of the protein sequence according to NP_064387.1.
Molecular Weight of Antigen 43.8 kDa
Purification Method Antigen Affinity-purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 112770.0
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.