Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCX1/SLC8A1 Polyclonal Antibody, Bioss
SDP

Catalog No. 502280924
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0924 Supplier Bioss Supplier No. BS1550R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NCX1/SLC8A1 Polyclonal antibody specifically detects NCX1/SLC8A1 in Human, Mouse, Rat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen NCX1/SLC8A1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene Slc8a1
Gene Accession No. P32418, P70414, Q01728
Gene Alias AI852629; AV344025; CNC; D930008O12Rik; Na(+)/Ca(2+)-exchange protein 1; Na/Ca exchanger; Na+/Ca++ exchanger; Na+/Ca2+ exchanger; Na-Ca exchanger; Ncx; Ncx1; Slc8a1; Sodium/calcium exchanger 1; sodium/calcium exchanger isoform NaCa7; solute carrier family 8 (sodium/calcium exchanger), member 1; solute carrier family 8 member 1; solute carrier family 8 member A1; solute carrier family 8, member 1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NCX1
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20541, 29715, 6546
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.