Learn More
ND1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 197031AP150UL
Description
This antibody detects both isoforms of ND1, 318aa 34-36 kDa and 268aa, 28-30 kDa.
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7),while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 1 (ND1) binds rotenone and rotenone analogs and might be involved in electron transfer to ubiquinone. Mutations in the ND1 gene may be implicated in several disorders, including Leber hereditary optic neuropathy, Alzheimer′s disease and Parkinson disease.Specifications
| ND1 | |
| Polyclonal | |
| Unconjugated | |
| MT-ND1 | |
| MT ND1, MTND1, NADH dehydrogenase subunit 1, NADH1, ND1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17716, 4535 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P03886 | |
| mt-Nd1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.