Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ND5 Mouse anti-Human, Clone: 1C12G8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | ND5 |
---|---|
Clone | 1C12G8 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7031
![]() ![]() |
Proteintech Group Inc
666131IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encodiing for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.Specifications
ND5 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
MT ND5, MTND5, NADH dehydrogenase subunit 5, NADH5, ND5 | |
MT-ND5 | |
IgG2b | |
Protein A | |
Antibody |
1C12G8 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P03915 | |
4540 | |
Peptide | |
Primary | |
-20°C | |
MT-ND5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title