Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDE1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106537DL594
Description
NDE1 Polyclonal antibody specifically detects NDE1 in Human samples. It is validated for Western BlotSpecifications
NDE1 | |
Polyclonal | |
Western Blot | |
FLJ20101, HOM-TES-87, nuclear distribution protein nudE homolog 1, NudE, nudE nuclear distribution gene E homolog 1 (A. nidulans), NUDE1, rat homolog | |
The immunogen recognized by this antibody maps to a region between residue 275 and 325 of human nuclear distribution gene E using the numbering given in entry NP_060138.1 (GeneID 54820). | |
0.1 mL | |
Cell Cycle and Replication | |
54820 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction