Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531121 Supplier Invitrogen™ Supplier No. PA531121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (80%), Xenopus laevis (82%), Dog (99%), Chicken (90%), Rhesus Monkey (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NDE1 is s a member of the nuclear distribution E (NudE) family of proteins. It is localized at the centrosome and interacts with other centrosome components as part of a multiprotein complex that regulates dynein function. NDE1 protein plays an essential role in microtubule organization, mitosis and neuronal migration. Mutations to the gene lead to Lissencephaly 4 (LIS4) and Microhydranencephaly (MHAC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDE1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene NDE1
Gene Accession No. Q9NXR1
Gene Alias 2810027M15Rik; AU042936; AW822251; HOM-TES-87; LIS1-interacting protein NUDE1; LIS1-interacting protein NUDE1, rat homolog; LIS4; MHAC; mNudE; NDE; NDE1; nuclear distribution gene E homolog 1; nuclear distribution gene E homolog 1 (A nidulans); nuclear distribution protein nudE homolog 1; NudE; nudE neurodevelopment protein 1; nudE nuclear distribution E homolog 1; nudE nuclear distribution gene E homolog 1; Nude1; rNudE
Gene Symbols NDE1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NDE1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54820
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.