Learn More
Invitrogen™ NDE1 Recombinant Superclonal™ Antibody
Rabbit Recombinant Superclonal Antibody
Supplier: Invitrogen™ 711424
Description
This antibody is predicted to react with Monkey, Cat, and Horse. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
NDE1 is s a member of the nuclear distribution E (NudE) family of proteins. It is localized at the centrosome and interacts with other centrosome components as part of a multiprotein complex that regulates dynein function. NDE1 protein plays an essential role in microtubule organization, mitosis and neuronal migration. Mutations to the gene lead to Lissencephaly 4 (LIS4) and Microhydranencephaly (MHAC).
Specifications
NDE1 | |
Recombinant Superclonal | |
Unconjugated | |
NDE1 | |
2810027M15Rik; AU042936; AW822251; HOM-TES-87; LIS1-interacting protein NUDE1; LIS1-interacting protein NUDE1, rat homolog; LIS4; MHAC; mNudE; NDE; NDE1; nuclear distribution gene E homolog 1; nuclear distribution gene E homolog 1 (A nidulans); nuclear distribution protein nudE homolog 1; NudE; nudE neurodevelopment protein 1; nudE nuclear distribution E homolog 1; nudE nuclear distribution gene E homolog 1; Nude1; rNudE | |
Rabbit | |
Protein A | |
RUO | |
54820, 67203, 83836 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9CZA6, Q9ES39, Q9NXR1 | |
NDE1 | |
Protein corresponding to human NDE1 (aa1-aa346). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.