Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDEL1 Monoclonal Antibody (OTI5C2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503313
Description
This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| NDEL1 | |
| Monoclonal | |
| Unconjugated | |
| NDEL1 | |
| EOPA, MITAP1, NDE1L1, NDE2, NUDEL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 81565 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5C2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9GZM8 | |
| NDEL1 | |
| Full length human recombit protein of human NDEL1 produced in HEK293 cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction