Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine, 98%, Thermo Scientific™

Catalog No. AAH6392503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6392503 1 g
AAH6392506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6392503 Supplier Thermo Scientific Chemicals Supplier No. H6392503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16937-92-1
Molecular Formula C18H26N2O6
Molecular Weight (g/mol) 366.41
MDL Number MFCD00069970
InChI Key QYYCZJUFHDLLOJ-UHFFFAOYNA-N
Synonym boc-d-orn z-oh, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, a-z-n, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid
PubChem CID 7272030
SMILES CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Beilstein 2400891
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
Quantity 1 g
Formula Weight 366.41
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.