Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDFIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569603 Supplier Invitrogen™ Supplier No. PA569603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

Activates HECT domain-containing E3 ubiquitin-protein ligases, including ITCH, NEDD4, NEDD4L, SMURF2, WWP1 and WWP2, and consequently modulates the stability of their targets. As a result, may control many cellular processes. Recruits ITCH, NEDD4 and SMURF2 to endosomal membranes. Negatively regulates KCNH2 potassium channel activity by decreasing its cell-surface expression and interfering with channel maturation through recruitment of NEDD4L to the Golgi apparatus and multivesicular body where it mediates KCNH2 degradation (PubMed:26363003). May modulate EGFR signaling. Together with NDFIP1, limits the cytokine signaling and expansion of effector Th2 T-cells by promoting degradation of JAK1, probably by ITCH- and NEDD4L-mediated ubiquitination. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDFIP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NDFIP2
Gene Accession No. 0
Gene Alias 0710001O20Rik; 2810436B12Rik; 9130207N19Rik; KIAA1165; MAPK-activating protein PM04 PM05 PM06 PM07; mKIAA1165; N4wbp5a; NDFIP2; Nedd4 family interacting protein 2; NEDD4 family-interacting protein 2; NEDD4 WW domain-binding protein 5A; NF-kappa-B-activating protein 413; putative MAPK-activating protein PM04/PM05/PM06/PM07; Putative NF-kappa-B-activating protein 413; Unknown (protein for IMAGE:7958216); wu:fa92h03; wu:fk31f07; zgc:63948
Gene Symbols NDFIP2
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat NDFIP2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361089
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.