Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDM-1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177688AF594
Description
NDM-1 Polyclonal specifically detects NDM-1 in Mouse, Bacteria, E. coli samples. It is validated for Western Blot.Specifications
NDM-1 | |
Polyclonal | |
Western Blot | |
Rabbit | |
Affinity Purified | |
Primary | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873] | |
0.1 mL | |
Mouse, Bacteria, E. coli | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction