Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDM-1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177688H
Description
NDM-1 Polyclonal specifically detects NDM-1 in Mouse, Bacteria, E. coli samples. It is validated for Western Blot.Specifications
| NDM-1 | |
| Polyclonal | |
| Western Blot | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Mouse, Bacteria, E. coli | |
| IgG |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873] | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction