Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDNL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550619 Supplier Invitrogen™ Supplier No. PA550619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total HCA-4 protein.

The protein encoded by this gene is part of the SMC5-6 chromatin reorganizing complex and is a member of the MAGE superfamily. This is an intronless gene. Component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination in ALT (alternative lengthening of telomeres) cell lines and mediates sumoylation of shelterin complex (telosome) components which is proposed to lead to shelterin complex disassembly in ALT-associated PML bodies (APBs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDNL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NSMCE3
Gene Accession No. Q96MG7, Q9CPR8
Gene Alias 5730494G16Rik; AI642138; BB044375; HCA4; Hepatocellular carcinoma-associated protein 4; hepatocellular carcinoma-associated protein HCA4; Mageg1; mage-g1; MAGE-G1 antigen; MAGEL3; magg1; melanoma antigen, family G, 1; melanoma-associated antigen G1; NDNL2; necdin-like 2; necdin-like 2, SMC5-SMC6 complex component; necdin-like gene 2; necdin-like protein 2; Non-SMC element 3 homolog; non-structural maintenance of chromosomes element 3 homolog; NSE3; NSE3 homolog, SMC5-SMC6 complex component; NSMCE3
Gene Symbols NSMCE3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human necdin-like 2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56160, 66647
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.