Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDOR1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298487MFV450
Description
NDOR1 Polyclonal antibody specifically detects NDOR1 in Human samples. It is validated for Western BlotSpecifications
NDOR1 | |
Polyclonal | |
Western Blot | |
bA350O14.9, EC 1.6, MGC138148, NADPH dependent diflavin oxidoreductase 1, NADPH-dependent diflavin oxidoreductase 1, NADPH-dependent FMN and FAD containing oxidoreductase, NADPH-dependent FMN and FAD-containing oxidoreductase, Novel reductase 1, NR1NADPH dependent FMN and FAD containing oxidoreductase | |
Produced in rabbits immunized with E. coli-derived Human NDOR1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
27158 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction