Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547431
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547431 Supplier Invitrogen™ Supplier No. PA547431
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Norrin is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDP
Applications Immunohistochemistry, Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene NDP
Gene Accession No. Q00604
Gene Alias EVR2; FEVR; ND; NDP; NDP, norrin cystine knot growth factor; Ndph; Norrie disease (pseudoglioma); Norrie disease (pseudoglioma) (human); Norrie disease homolog; norrie disease protein; Norrie disease protein homolog; norrin; norrin cystine knot growth factor NDP; RGD1563968; similar to norrin; X-linked exudative vitreoretinopathy 2 protein
Gene Symbols NDP
Host Species Goat
Immunogen E. coli-derived recombinant human Norrin Lys25-Ser133.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4693
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.