Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582059 Supplier Invitrogen™ Supplier No. PA582059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDSSFIMDSD PRRCMRHHYV DSISHPLYKC SSKMVLLARC EGHCSQASRS EPLVSFSTVL KQPFRSSCHC CRPQTSKLKA LRLRCSGGMR LTATYRYILS CHCEECNS Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.

ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDP
Gene Accession No. Q00604
Gene Alias EVR2; FEVR; ND; NDP; NDP, norrin cystine knot growth factor; Ndph; Norrie disease (pseudoglioma); Norrie disease (pseudoglioma) (human); Norrie disease homolog; norrie disease protein; Norrie disease protein homolog; norrin; norrin cystine knot growth factor NDP; RGD1563968; similar to norrin; X-linked exudative vitreoretinopathy 2 protein
Gene Symbols NDP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDP. Recombinant protein control fragment (Product #RP-92199).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4693
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.