Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501730190 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0190 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0190 Supplier Proteintech Group Inc Supplier No. 153801AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects 11-15 kDa (monomer) and 48 kDa (polymers).

Norrie disease is an X-linked genetic disorder characterized by progressive atrophy of the eyes, mental disturbances and deafness. The gene responsible for this disease was initially identified through positional cloning. Norrin, the gene product, encodes a small secreted, cysteine-rich protein that is thought to act as a ligand for the Wnt-receptor/beta-catenin signal pathway despite having sequence homology with the Wnt family of proteins. Mice lacking this gene have abnormal blood vessel growth in the vitreous and a disorganized retina, transgenic ectopic expression of Norrin restores normal retinal vasculature. Recent evidence shows that Norrin can attenuate tPA and uPA-mediated death of transformed rat retinal ganglion cells (RGC-5) by activating the Wnt/beta-catenin pathway and regulating the phosphorylation of LRP-1, a cell surface receptor for tPA and uPA, suggesting the Norrin may function in vivo by regulating kinases which may alter the phosphorylation of LRP-1.

Specifications

Antigen NDP
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NDP
Gene Accession No. P48744, Q00604
Gene Alias EVR2, FEVR, ND, NDP, Norrie disease (pseudoglioma), Norrie disease protein, Norrin
Gene Symbols Ndp
Host Species Rabbit
Immunogen NDP Fusion Protein Ag4066
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17986, 4693
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.