Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDP52 Antibody (OTI4H5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272897FR
Description
NDP52 Monoclonal specifically detects NDP52 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
NDP52 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
CALCOCO2 | |
Full length recombinant protein of human NDP52 (NP_005822) produced in HEK293T cells. | |
0.1 mL | |
10241.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI4H5 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Antigen nuclear dot 52 kDa protein, calcium binding and coiled-coil domain 2, calcium-binding and coiled-coil domain-containing protein 2, NDP52MGC17318, Nuclear domain 10 protein 52, Nuclear domain 10 protein NDP52, Nuclear dot protein 52 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction